missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oleoyl-L-alpha-lysophosphatidic acid sodium salt

Product Code. 15408359
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408359 5 mg 5mg
15418359 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408359 Supplier Thermo Scientific Chemicals Supplier No. J66836.#0
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22556-62-3 | C21H40NaO7P, C21H40NaO7P | 458.51 g/mol

It is an endogenous agonist for LPA1 and LPA2 receptors. LPA does not induce angiogenesis, but has effects on endothelial cell physiology that are similar to those of sphingosine 1-phosphate. Induces cell migration of cancer and non-cancer cells. LPA has also been described to potentiate acetylchloine receptor currents by G-protein-mediated activation of Ca2+-dependent/independent protein kinase C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22556-62-3
Molecular Formula C21H40NaO7P
Synonym 1-O-9Z-Octadecenoyl-sn-glyceryl-3-phosphoric acid sodium salt; Lysophosphatidic Acid

Specifications

Quantity 5 mg
Description White to off-white
Content And Storage -30°C to -10°C
Formula Weight 458.5
Sensitivity Air and Light Sensitive, Hygroscopic
Solubility Soluble in water up to 5mg/ml with sonication
Product Type Oleoyl-L-alpha-lysophosphatidic acid sodium salt
Form Waxy solid or powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.