missing translation for 'onlineSavingsMsg'
Learn More

Oleyl alcohol, ca. 60%, technical

Product Code. 10091400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091400 1 L Glass Bottle 1 L
10360114 2.5 L Glass Bottle 2.5 L
10419250 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10091400 Supplier Thermo Scientific Acros Supplier No. 129530010
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 143-28-2 | C18H36O | 268.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-28-2
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.49
MDL Number MFCD00002993
InChI Key ALSTYHKOOCGGFT-KTKRTIGZSA-N
Synonym oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol
PubChem CID 5284499
ChEBI CHEBI:73504
IUPAC Name (9Z)-octadec-9-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCO

Specifications

Melting Point 5.0°C
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 207.0°C (13.0 mmHg)
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 60% Typical (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)7CH=CH(CH2)8OH
Refractive Index 1.4590 to 1.4640
Quantity 1 L
Beilstein 01, 453
Merck Index 15, 6924
Specific Gravity 0.84
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol and ether, soluble in warbontetrachloride
Formula Weight 268.47
Percent Purity ca. 60%
Grade Technical
Physical Form Viscous Liquid
Chemical Name or Material Oleyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • Oleyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.