missing translation for 'onlineSavingsMsg'
Learn More

Oleylamine, approximate C18-content 80-90%

Product Code. 10400380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
1 L
5 L
Unit Size:
1 L
100mL
5 L
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10400380 100 mL 100mL
10316630 5 mL 5mL
10356580 1 L 1 L
15254624 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10400380 Supplier Thermo Scientific Acros Supplier No. 129541000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112-90-3 | C18H37N | 267.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-90-3
Molecular Formula C18H37N
Molecular Weight (g/mol) 267.50
MDL Number MFCD00066507
InChI Key QGLWBTPVKHMVHM-KTKRTIGZSA-N
Synonym 9-octadecenylamine, 9-octadecen-1-amine, cis-9-octadecenylamine, e-octadec-9-en-1-amine, 1-amino-9-octadecene, 9e octadec-9-enylamine, 9-octadecenylamine 8ci, 36505-83-6 hydrofluoride, 3811-68-5 unspecified acetate
PubChem CID 6258392
SMILES CCCCCCCC\C=C/CCCCCCCCN

Specifications

Melting Point 15.0°C to 22.0°C
Color Yellow
Density 0.8100g/mL
Boiling Point 147.0°C (2.0 mmHg)
Flash Point 154°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Linear Formula CH3(CH2)7CH=CH(CH2)8NH2
Refractive Index 1.4585 to 1.4625 (20°C, 589nm)
Quantity 100 mL
UN Number 2735
IUPAC Name (E)-octadec-9-en-1-amine
Formula Weight 267.5
Percent Purity ≥96.0% (primary amine)
Physical Form Liquid After Melting
Chemical Name or Material Oleylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Oleylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.