missing translation for 'onlineSavingsMsg'
Learn More

Oleylamine, TRC

Product Code. 30409069
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409069 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409069 Supplier Toronto Research Chemicals Supplier No. TRCO532968100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Z)-9-Octadecenylamine
CAS 112-90-3
Molecular Formula C18 H37 N
Quantity 100 g
Synonym (9Z)-9-Octadecen-1-amine (ACI), 9-Octadecen-1-amine','(Z)- (ZCI), 9-Octadecenylamine','(Z)- (8CI), ((Z)-9-Octadecenyl)amine, 9-(Z)-Octadecen-1-amine, Adogen 172D, Alamine 11, Amine OB, Armeen O, Armeen OD, Armeen OLD-C, Armeen OM 97, Arosurf AA 60, cis-9-Oc
InChI Formula InChI=1S/C18H37N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10H,2-8,11-19H2,1H3/b10-9-
SMILES CCCCCCCC\C=C/CCCCCCCCN
IUPAC Name (Z)-octadec-9-en-1-amine
Molecular Weight (g/mol) 267.493
Formula Weight 267.293
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.