missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oligo(dT)18 Primer

Product Code. 10753741 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
60 μL
120 μL
Unit Size:
120µL
60µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10753741 120 μL 120µL
10304690 60 μL 60µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10753741 Supplier Thermo Scientific™ Supplier No. SO132
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends.

Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready-to-use, 20X concentrated aqueous solution.

Applications
• cDNA Synthesis
• RT-qPCR
• RT-PCR

Related products
Anchored Oligo dT (Cat. No. AB1247)
Random Hexamer Primer (Cat. No. SO142)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Primer
Content And Storage • Oligo(dT)18 Primer (120 μL at 0.5 μg/μL)

Store at –20°C.
Primer Length 18-mer
Shipping Condition Dry Ice
Concentration 0.5 μg/μL
Primer Oligo dT
Quantity 120 μL
For Use With (Application) cDNA Synthesis
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.