missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ OLIGO™ R3 Reversed - Phase Resin

Product Code. 18446788 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
25 mL
50 mL
1 L
Unit Size:
1 L
25mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18446788 50 mL 50mL
18416568 25 mL 25mL
18476728 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18446788 Supplier Thermo Scientific™ Supplier No. 1133906
  • ONLINE EXCLUSIVE PRICE
    1455.00 €
    1396.00 € / 50mL
    Save 59.00 €
    4% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules.

Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules. The combination of 30-μm particle size, resistance to high pH, and high-phase ratio, makes Oligo R3 resin particularly suited to preparative purification of synthetic oligonucleotides and peptides. The packing consists of cross-linked poly-divinylbenzene particles.

Oligo R3 bulk resin differs somewhat in selectivity from conventional silica-based, reversed-phase resins and is similar to high-carbon-loading through C18 supports. In addition, the complete lack of residual silanol activity may modify the selectivity of ionic species.

Features of Oligo R3 resin and pre-packed columns include:

  • High performance chromatography resin for analytical to process scale separations
  • High resolution preparative purification at high flow rates
  • Chemical stability—allows aggressive cleaning and sanitization
  • Poly-divinylbenzene particles—yield a robust, easily packable matrix
  • Combines low back pressure, high pH stability, and high capacity

Specifications

Matrix Cross-linked poly(styrene-divinylbenzene) POROS beads
Content And Storage Store at 2°C to 30°C. Do not freeze.
Shipping Condition Ambient
Particle Size 30 μm
Form Resin Slurry
Product Line OLIGO
Quantity 50 mL
Type Resin
Stationary Phase R3
Format Bulk Media
Binding Capacity 25 mg Lysozyme/mL Resin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.