missing translation for 'onlineSavingsMsg'
Learn More

Olive oil, pure, virgin

Product Code. 10667031 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
25mL
500mL
This item is not returnable. View return policy

Product Code. 10667031

Brand: Thermo Scientific Acros 416540250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8001-25-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-25-0

Specifications

Acidity 2.0mg KOH/g max.
Absorbance 0.2 (270nm, 1g/100ml cyclohexane) max. at 425nm
Color Yellow
Density 0.9135g/mL
Flash Point 225°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4670 to 1.4710
Quantity 25 mL
Saponification Value 184 to 196mg KOH/g
Merck Index 14,6836
Specific Gravity 0.9135
Solubility Information Solubility in water: .. Other solubilities: practically insoluble in alcohol, miscible with light petroleum (50-60°C)
Grade Pure
Concentration or Composition (by Analyte or Components) of fatty acids: 0.1 % max. (< C16) 7.5 to 20.0 % (Palmitic acid) 3.5 % max. (Palmitoleic acid) 0.5 to 5.0 % (Stearic acid) 56.0 to 85.0 % (Oleic acid) 3.5 to 20.0 % (Linoleic acid) 1.2 % max. (Linolenic acid) 0.7 % max. (Arachidic acid) 0.4 % max. (Eic
Physical Form Powder
Chemical Name or Material Olive oil
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.