missing translation for 'onlineSavingsMsg'
Learn More

Olodaterol Benzyl Ether, TRC

Product Code. 30415871
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415871 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415871 Supplier Toronto Research Chemicals Supplier No. TRCO26200510MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Olodaterol Benzyl Ether
CAS 869478-13-7
Molecular Formula C28H32N2O5
Quantity 10 mg
Synonym 8-[(1R)-1-Hydroxy-2-[[2-(4-methoxyphenyl)-1,1-dimethylethyl]amino]ethyl]-6-(phenylmethoxy)-2H-1,4-benzoxazin-3-(4H)-one
InChI Formula InChI=1S/C28H32N2O5/c1-28(2,15-19-9-11-21(33-3)12-10-19)29-16-25(31)23-13-22(34-17-20-7-5-4-6-8-20)14-24-27(23)35-18-26(32)30-24/h4-14,25,29,31H,15-18H2,1-3H3,(H,30,32)/t25-/m0/s1
SMILES COC(C=C1)=CC=C1CC(C)(C)NC[C@@H](C2=C3C(NC(CO3)=O)=CC(OCC4=CC=CC=C4)=C2)O
IUPAC Name (R)-6-(benzyloxy)-8-(1-hydroxy-2-((1-(4-methoxyphenyl)-2-methylpropan-2-yl)amino)ethyl)-2H-benzo[b][1,4]oxazin-3(4H)-one
Molecular Weight (g/mol) 476.56
Formula Weight 476.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.