missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Olopatadine hydrochloride
Description
Histamine H1 receptor antagonist (Ki = 31.6 nM). Inhibits the release of histamine, prostaglandin D2 and tryptase in a concentration-dependent manner. Mast cell stabilizer; inhibits mast cell mediator release. Also suppresses inflammation by inhibition of cytokine production.
Specifications
Specifications
| Quantity | 10 mg |
| Product Line | Tocris |
| Formula Weight | 373.87 |
| Purity | >99% |
| Molecular Weight (g/mol) | 373.877 |
| Chemical Name or Material | Olopatadine hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?