missing translation for 'onlineSavingsMsg'
Learn More

Olprinone Hydrochloride, MedChemExpress

Product Code. p-200101957
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16442147

Brand: Medchem Express HY14254

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Olprinone (Loprinone) Hydrochloride is a potent phosphodiesterase (PDE) 3 inhibitor, with IC50s of 150, 100, 0.35 and 14 μM for PDE1, PDE2, PDE3 and PDE4, respectively. Olprinone Hydrochloride is used for the research of heart failure due to its positive inotropic and vasodilative effects. Anti-inflammatory activity.

Olprinone (Loprinone) Hydrochloride is a potent phosphodiesterase (PDE) 3 inhibitor, with IC50s of 150, 100, 0.35 and 14 μM for PDE1, PDE2, PDE3 and PDE4, respectively. Olprinone Hydrochloride is used for the research of heart failure due to its positive inotropic and vasodilative effects. Anti-inflammatory activity.

Specifications

Chemical Name or Material Olprinone Hydrochloride
CAS 119615-63-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H11ClN4O
Quantity 50 mg
Synonym Loprinone Hydrochloride
Solubility Information H2O : ≥ 7.69 mg/mL (26.82 mM)
SMILES CC(N1)=C(C=C(C#N)C1=O)C2=CN3C(C=C2)=NC=C3.[H]Cl
Molecular Weight (g/mol) 286.72
Formula Weight 286.72
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.