Learn More
EVOS™ Olympus 4X Objective, super-apochromat, LWD
Product Code. 15691882
Change view
Click to view available options
Magnification Power:
4X
Unit Size:
Each
Product Code. Magnification Power unitSize
15691882 4X Each
1 options
Product Code. 15691882 Supplier EVOS™ Supplier No. AMEP4752
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for demanding research applications
  • Has long working distance (LWD) objective that is optimized for imaging of slides, cell culture dishes and flasks, and microtiter plates
  • Offers Apo correction from UV through IR ranges
  • Parfocal for both visible and IR imaging
  • Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
  • Compatible with EVOS Cell Imaging systems
  • Offers optical performance from visible light to near infrared light
  • Meets needs across spectrum of magnifications and optical specifications
TRUSTED_SUSTAINABILITY
Specifications
Type Lens
Product Line EVOS
Focus Distance 13 mm
For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Includes Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
Lens Type Super-apochromat
Magnification Power 4X
Numerical Aperture 0.16
Quantity Each
Shipping Condition Room Temperature