Learn More
Thermo Scientific™ Olympus™ 4X Objective, U Plan S-Apo, 0.16 NA
Product Code. 15271178 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. Quantity Unit Size
15271178 1 each Each
1 options
Product Code. 15271178 Supplier Thermo Scientific™ Supplier No. NX1004X
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
TRUSTED_SUSTAINABILITY
Specifications
Type Objective Lens
Product Line Olympus
Description Olympus™ 4X Objective, U Plan S-Apo (0.16 NA)
For Use With (Equipment) CellInsight CX5 HCS Platform
Lens Type Dry
Magnification 4x (0.16NA)
Quantity 1 each