missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole metabolite Omeprazole sulfone, MedChemExpress

Product Code. 16468788
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468788 10 mg 10mg
16461218 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468788 Supplier Medchem Express Supplier No. HYG0007
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Omeprazole sulfone is a metabolite of Omeprazole, which is a proton pump inhibitor.

Omeprazole sulfone is a metabolite of Omeprazole, which is a proton pump inhibitor.

Specifications

Chemical Name or Material Omeprazole metabolite Omeprazole sulfone
CAS 88546-55-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H19N3O4S
Quantity 10 mg
Synonym Omeprazole sulfone Omeprazole sulphone
SMILES CC(C(OC)=C(C)C=N1)=C1CS(C2=NC3=CC=C(OC)C=C3N2)(=O)=O
Molecular Weight (g/mol) 361.42
Formula Weight 361.42
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.