missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole N-Oxide, TRC

Product Code. 17995005
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17995005 50 mg 50mg
17932545 5 mg 5mg
17981025 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17995005 Supplier Toronto Research Chemicals Supplier No. TRCO63501550MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176219-04-8
Molecular Formula C17 H19 N3 O4 S
Molecular Weight (g/mol) 361.42
Synonym 4-Methoxy-2-[[(RS)-(5-methoxy-1H-benzimidazol-2-yl)sulphinyl]methyl]-3,5-dimethylpyridine 1-Oxide, Esomeprazole Magnesium Trihydrate Imp. E (EP), Omeprazole Imp. E (EP), Omeprazole N-Oxide
IUPAC Name 5-methoxy-2-[(4-methoxy-3,5-dimethyl-1-oxidopyridin-1-ium-2-yl)methylsulfinyl]-1H-benzimidazole
SMILES COc1ccc2[nH]c(nc2c1)S(=O)Cc3c(C)c(OC)c(C)c[n+]3[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H19N3O4S/c1-10-8-20(21)15(11(2)16(10)24-4)9-25(22)17-18-13-6-5-12(23-3)7-14(13)19-17/h5-8H,9H2,1-4H3,(H,18,19)
Formula Weight 361.1096
Percent Purity >95%
Chemical Name or Material Omeprazole N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.