missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole, TRC

Product Code. 17924855
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17924855 100 mg 100mg
17901035 50 mg 50mg
17912045 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17924855 Supplier Toronto Research Chemicals Supplier No. TRCO635000100MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73590-58-6
Molecular Formula C17 H19 N3 O3 S
Molecular Weight (g/mol) 345.42
Synonym 1H-Benzimidazole, 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]- (9CI), (±)-Omeprazole, 2-[[(3,5-Dimethyl-4-methoxy-2-pyridyl)methyl]sulfinyl]-5-methoxy-1H-benzimidazole, 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridyl)methyl]sulfinyl]-1H-benzimidazole, Acidex, Antra, Antra MUPS, Audazol, Aulcer, Belmazol, Ceprandal, Desec, Dizprazol, Dudencer, Elgam, Emeproton, Epirazole, Gastrimut, GastroGard, Gastroloc, Gastrozole, Gibancer, H 168/68, Indurgan, Inhibitron, Inhipump, Logastric, Lomac, Losec, Mepral, Miol, Miracid, Mopral, OMEP, OMP, OMZ, Ocid, Omapren, Omebeta 20, Omed, Omedar, Omepradex, Omepral, Omeprazen, Omeprazole, Omeprazon, Omepril, Omezol, Omezzol, Omid, Omisec, Omizac, Ompanyt, Oprax, Opraz, Ozoken, Parizac, Pepticum, Peptilcer, Prazidec, Prilosec, Prysma, Ramezol, Roweprazol, Sanamidol, Secrepina, Ulceral, Ulcesep, Ulcometion, Ulcozol, Ulsen, Ultop, Ulzone, Zefxon, Zimor, Zoltum
IUPAC Name 5-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
SMILES COc1ccc2[nH]c(nc2c1)S(=O)Cc3ncc(C)c(OC)c3C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H19N3O3S/c1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)
Formula Weight 345.1147
Percent Purity >95%
Chemical Name or Material Omeprazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.