missing translation for 'onlineSavingsMsg'
Learn More

BD™ OMICS-Guard Sample Preservation Buffer Kit

Product Code. 32188592
Change view
Click to view available options
Quantity:
12 x 1 mL
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32188592 12 x 1 mL 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32188592 Supplier BD™ Supplier No. 570908
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The BD™ OMICS-Guard Sample Preservation Buffer is developed and optimized to address the need for high-quality biological sample preservation over an extended period of time when samples cannot be processed at the same time or location.

  • Provides a stress-free, one-step preservation protocol with minimum hands-on time
  • Optimized to preserve cells for a variety of downstream transcriptomic, proteomic and multiomic applications, including RNA-seq, CITE-seq, flow cytometry and qPCR
  • Protects cell viability and preserves different cell populations in your samples for up to 72 hours at 4°C
  • Developed and tested across multiple sample types: PBMCs and tissue samples
  • Available in 12 x 1 mL vials
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) BD Rhapsody platform, Single-Cell Analysis System
Product Type Sample Preservation Buffer
Format Vial
Quantity 12 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.