Learn More
B Braun™ Ominifix™ Solo Luer Lock Cone
Product Code. 12702687
Change view
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Pack of 100
Product Code. Volume (Metric) Unit Size
12702687 50 mL Pack of 100
1 options
Product Code. 12702687 Supplier B Braun™ Supplier No. 4617509F
  • ONLINE EXCLUSIVE PRICE
    192.00 €
    155.92 € / Pack of 100
    Save 36.08 €
    18% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Certifications/Compliance EN ISO 7886-1
Material Polypropylene
Product Type Centric, Luer Lock
Disposable Single-use
Quantity 1/Ea.
Volume (Metric) 50 mL
Sterility Sterile