missing translation for 'onlineSavingsMsg'
Learn More

Ion Torrent™ Oncomine™ Comprehensive Assay Plus GX

Product Code. 31034206
Change view
Click to view available options
Quantity:
16 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31034206 16 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31034206 Supplier Ion Torrent™ Supplier No. A47013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Oncomine Comprehensive Assay Plus GX is a targeted next-generation sequencing (NGS) assay that provides a comprehensive genomic profiling (CGP) solution appropriate for formalin-fixed paraffin-embedded (FFPE) tissues from solid tumors.

The Ion Torrent Oncomine Comprehensive Assay Plus GX is a targeted next-generation sequencing (NGS) assay that provides a comprehensive genomic profiling (CGP) solution appropriate for formalin-fixed paraffin-embedded (FFPE) tissues from solid tumors. The assay allows concurrent analysis of DNA and RNA to simultaneously detect a broad range of genomic alterations from 517 genes, including single-nucleotide variants (SNVs), insertions and deletions (indels), copy number variations (CNVs), and fusions. Additionally, the assay detects genomic signatures such as homologous recombination deficiency (HRD) using genomic instability metric (GIM), tumor mutational burden (TMB), and microsatellite instability (MSI).

Key features of the assay include:

  • Enables CGP across 517 genes
  • Accurately detects SNVs, indels, CNVs, including large genomic rearrangements (LGRs) in BRCA1 and BRCA2, and fusions including splice variants
  • Assesses for genomic signatures, including HRD, TMB, and MSI, to help maximize insights on the underlying oncogenic drivers
  • Provides robust performance from 30 ng of DNA and/or 20 ng of RNA isolated from FFPE samples, including fine needle biopsies
  • Provides multiplex sequencing of up to four DNA samples (DNA + RNA or DNA-only) with optional NTC on a GX7 Chip in a single run
  • Provides multiplex sequencing of up to eight RNA-only samples (or eight plus optional NTC) per GX7
  • System-installed assays preconfigured with settings optimized for specific applications and sample types
  • Requires as little as 10 minutes hands-on time per run for easy, automated workflows
  • Provides automated Nucleic Acid to Result workflow on the Genexus Integrated Sequencer with next-day results

When used with the Genexus Purification System and the Genexus Integrated Sequencer, sequential sample purification, library preparation, sequencing, and analysis allow for the reporting of next-day results. With just two user touchpoints and as little as 20 minutes of hands-on time, the easy, automated workflows can help reduce handling errors, free up time for your laboratory staff, and reduce labor costs. As a single vendor of sample-to-report solutions, including instruments, consumables, analysis, and support, Thermo Fisher Scientific simplifies bringing in-house CGP to your lab at varying levels of NGS expertise.

TRUSTED_SUSTAINABILITY

Specifications

Sample Type DNA and RNA
Sequencing Type Next Generation Sequencing
For Use With (Equipment) Genexus™ Integrated Sequencer
Product Type Oncomine Comprehensive Assay Plus GX
Quantity 16 Samples

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.