missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oncomine™ Tumor Mutation Load Assay, manual library preparation
Specifications
Specifications
| Content And Storage | • 1 Oncomine Tumor Mutation Load Panel, 24 reactions, store at -5 to -30°C. • 1 Ion AmpliSeq Library Kit Plus, 24 reactions, store at -5 to -30°C. |
| Sequencing Type | Genome & DNA Sequencing |
| For Use With (Equipment) | Ion GeneStudio S5 System, Ion S5™ XL System |
| Libraries | Targeted Sequencing Library |
| Product Line | Oncomine |
| Product Type | Tumor Mutation Load Assay |
| Quantity | 24 samples |
| Shipping Condition | Dry Ice |
| Throughput | 24 Samples |
| Workflow Step | Library Generation |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?