missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ OneStep Blocker Western Blocking Solution & Signal Enhancer

Product Code. 30161300
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30161300 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30161300 Supplier Novus Biologicals™ Supplier No. NBP333180
  • ONLINE EXCLUSIVE PRICE
    115.00 €
    108.15 € / 500mL
    Save 6.85 €
    5% Off
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

provides blocking and primary and secondary antibody hybridization in one step and enhances the signal developed with HRP

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.