missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ONO 6818

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
ONO 6818 is a high affinity and selective human neutrophil elastase 1 (HNE1) inhibitor (Ki = 12 nM). Exhibits >100-fold less activity at other related proteases including trypsin, pancreatic elastase, collagenase and murine macrophage elastase. Inhibits HNE-induced lung hemorrhaging and suppresses leukocyte levels in an in vivo model of COPD. Orally active.
Specifications
Specifications
| Chemical Name or Material | 5-Amino-N-[1-[[5-(1,1-dimethylethyl)-1,3,4-oxadiazol-2-yl]carbonyl]-2-methylpropyl]-6-oxo-2-phenyl-1(6H)-pyrimidineacetamide |
| CAS | 208848-19-5 |
| Quantity | 10 mg |
| Target | Synthetic Elastase Inhibitors |
| Molecular Formula | C23H28N6O4 |
| Formula Weight | 452.51 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction