missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ONO AE3 208

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ONO AE3 208 is a high affinity and selective EP4 receptor antagonist (Ki values are 1.3, 30, 790 and 2400 nM for EP4, EP3, FP and TP receptors respectively). Displays no affinity for EP1, EP2, DP or IP receptors (Ki >10 μM). Inhibits PGE2-induced IL-8 production in colonic epithelial caco-2 cells and attenuates PGE2 inhibition of natural killer T cell activation. Suppresses recovery from experimentally-induced colitis and stimulates CD4+ T cell proliferation in C57BL/6 mice. Also reduces metastasis of mammary tumor cells in a murine model of breast cancer. Orally active.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | ONO AE3 208 |
| Purity | >98% |
| Molecular Weight (g/mol) | 404.43 |
| Inhibitors | ONO AE3 208 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction