Learn More
Kolmi Hopen™ Op-Air™ One Anti-glare Medical Mask with Foam Band
Be prepared for long hours of wear with this anti-glare, anti-fog medical mask complete with a comfortable foam band and anti-reflective tape.
Brand:  Kolmi Hopen™ M24101MN-30
Additional Details : Weight : 3.42000kg
Product Code. 17740189
  • 9.45 € / Pack of 50
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
  • High filtration media (polypropylene)
  • Layers (interior to exterior): wet laid cellulose, melt blown polypropylene, spunbonded polypropylene
  • Soft, non-woven mask ties and edges
  • Anatomical folding limits exposure to face
  • Integrated nose bar and foam strip to reduce glasses fogging
  • Black, anti-reflective tape limits glare phenomena in operating room
  • Fiberglass-free filter
  • Produced in ISO 8 clean room (particle and microbiological control)
Specifications
Specifications
Blue
Polypropylene
Yes
50 Pk.
Ties
(EU) 2017/745, EN 14683, ISO 13485, ISO 9001
Medical