missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ OpenArray™ 384-well Sample Plates

Product Code. 15661772
Change view
Click to view available options
Quantity:
10 plates
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15661772 10 plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15661772 Supplier Applied Biosystems™ Supplier No. 4406947
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with the QuantStudio™ 12K Flex Real-Time PCR System.

They are low volume microplates used in the QuantStudio™ OpenArray™ AccuFill™ System for sample loading onto OpenArray™ plates.

4406947
Color: Clear, Reaction Speed: Standard

NC0973054
Color: Black, Reaction Speed: Standard

TRUSTED_SUSTAINABILITY

Specifications

Format 384-well Plate
Content And Storage Each box contains 10 x 384-well plates, enough for 80 OpenArray™ Real-Time PCR plates.
For Use With (Equipment) QuantStudio™ 12 K Flex
Reaction Speed Standard
Product Line OpenArray
Product Type Sample Plate
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Room Temperature
Description Clear
Quantity 10 plates
Barcode No Barcode
Color Clear
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.