missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ OpenArray™ AccuFill™ System Tips

Product Code. 15685315 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Box
10 Boxes
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15685315 10 Boxes Pack of 10
15662222 1 Box Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15685315 Supplier Applied Biosystems™ Supplier No. 4458107
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This is a box of tips for use with the OpenArray™ AccuFill™ System, which is part of the OpenArray™ Real-Time PCR Platform.

Since AccuFill tips have a shorter shelf-life than OpenArray plates, we recommend that you align your order quantity with your estimated OpenArray throughput.About the OpenArray Real-Time PCR Platform:The OpenArray Real-Time PCR Platform provides PCR-based genotyping and gene expression analysis of hundreds to thousands of samples and assays per day. Researchers can analyze up to 144 samples on a single OpenArray plate. Three OpenArray plates can be cycled and imaged simultaneously on the OpenArray™ Real-Time PCR Instrument, providing high-throughput results with ease. The system workflow thermal cycles and fluorescently images the OpenArray plates. Results are captured on the external PC, and the OpenArray analysis software enables researchers to view specific assay data.

Since AccuFill tips have a shorter shelf-life than OpenArray plates, we recommend that you align your order quantity with your estimated OpenArray throughput.

About the OpenArray Real-Time PCR Platform
The OpenArray Real-Time PCR Platform provides PCR-based genotyping and gene expression analysis of hundreds to thousands of samples and assays per day. Researchers can analyze up to 144 samples on a single OpenArray plate. Three OpenArray plates can be cycled and imaged simultaneously on the OpenArray™ Real-Time PCR Instrument, providing high-throughput results with ease. The system workflow thermal cycles and fluorescently images the OpenArray plates. Results are captured on the external PC, and the OpenArray analysis software enables researchers to view specific assay data.

Complete Platform
The OpenArray Real-Time PCR Platform includes:
• OpenArray Real-Time PCR Instrument—images three OpenArray plates simultaneously
• OpenArray AccuFill System—loads samples into OpenArray plates
• OpenArray™ Case Sealing Station—seals OpenArray plates
• OpenArray instrument accessories (includes PC system)
• OpenArray™ Genotyping Analysis Software
• OpenArray™ Real-Time PCR Analysis Software

TRUSTED_SUSTAINABILITY

Specifications

Format Tips
Content And Storage Each Pk. contains 10 Boxes of AccuFill™ tips, with 384 tips / box.
For Use With (Equipment) OpenArray™
Product Line AccuFill, OpenArray
Product Type System Tip
For Use With (Application) Real Time PCR (qPCR)
Quantity 10 Boxes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.