missing translation for 'onlineSavingsMsg'
Learn More

Ophthazin Sulfate, TRC

Product Code. 30138163
Change view
Click to view available options
Quantity:
2.5 mg
100 mg
5 g
Unit Size:
100mg
2.5mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138163 2.5 mg 2.5mg
30136179 5 g 5g
30008386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138163 Supplier Toronto Research Chemicals Supplier No. TRCO6692002.5MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7327-87-9
Molecular Formula C8 H10 N6 . H2 O4 S
Molecular Weight (g/mol) 288.28
Synonym Phthalazine, 1,4-dihydrazinyl-, sulfate (1:1), 1,4-Phthalazinedione, 2,3-dihydro-, dihydrazone, sulfate (1:1) (9CI), Phthalazine, 1,4-dihydrazino-, sulfate (1:1) (8CI), 1,4-Dihydrazinophthalazine sulfate, Depressan, Dihydralazine sulfate, Dihydrazinophthalazine sulfate, Dihyzin
IUPAC Name (Z)-[(4Z)-4-hydrazinylidene-2,3-dihydrophthalazin-1-ylidene]hydrazine;sulfuric acid
SMILES N\N=C\1/NN\C(=N/N)\c2ccccc12.OS(=O)(=O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C8H10N6.H2O4S/c9-11-7-5-3-1-2-4-6(5)8(12-10)14-13-7;1-5(2,3)4/h1-4H,9-10H2,(H,11,13)(H,12,14);(H2,1,2,3,4)
Formula Weight 288.0641
Chemical Name or Material Dihydralazine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.