missing translation for 'onlineSavingsMsg'
Learn More

Opipramol, TRC

Product Code. 17930615
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17930615 10 mg 10mg
30137248 25 mg 25mg
17901635 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17930615 Supplier Toronto Research Chemicals Supplier No. TRCO66960010MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 315-72-0
Molecular Formula C23 H29 N3 O
Molecular Weight (g/mol) 363.5
Synonym 1-Piperazineethanol, 4-[3-(5H-dibenz[b,f]azepin-5-yl)propyl]-, 5H-Dibenz[b,f]azepine, 1-piperazineethanol deriv., 4-[3-(5H-Dibenz[b,f]azepin-5-yl)propyl]-1-piperazineethanol, Endison, G 33040, GR 33040, N-[3-[4-(2-Hydroxyethyl)piperazino]propyl]iminostilbene, NSC 169867, Opipramol, Opipramol G, Opramidol
IUPAC Name 2-[4-(3-benzo[b][1]benzazepin-11-ylpropyl)piperazin-1-yl]ethanol
SMILES OCCN1CCN(CCCN2c3ccccc3C=Cc4ccccc24)CC1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H29N3O/c27-19-18-25-16-14-24(15-17-25)12-5-13-26-22-8-3-1-6-20(22)10-11-21-7-2-4-9-23(21)26/h1-4,6-11,27H,5,12-19H2
Formula Weight 363.2311
Percent Purity >95%
Chemical Name or Material Opipramol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.