missing translation for 'onlineSavingsMsg'
Learn More

Chemtronics Optic Prep™ Highest Quality Premoistened Lens-Grade Tissue for Cleaning Optical Surfaces

Product Code. 17210592 Shop All Texwipe Products
Change view
Click to view available options
Color-coded:
No
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color-coded Unit Size
17210592 No Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17210592 Supplier Chemtronics Supplier No. CP410
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Presaturated Wipes are high quality premoistened lens-grade tissues for cleaning optical surfaces.

Optic Prep™ Presaturated Wipes contain ultra-pure isopropyl alcohol (65% IPA / 35% Pure Water) on an ultra-soft, lens-grade tissue.

Features and Benefits

  • Safely cleans and polishes optical surfaces, lenses, photographic equipment, LCD and CRT screens and instruments
  • Evaporates quickly, leaves no residue
  • Compatible with most plastics, glass and metals
  • Mild nonabrasive solvent
  • Quickly removes dust, oils and contaminants

Applications

  • Safely cleans and polishes optical surfaces, lenses, photographic equipment, fiber optic connectors and instruments
  • Innovative formula gently cleans LCD and plasma screens

Specifications

Product Type Lens Cleaning Tissue
Color-coded No
Form Sachet Wipe
Scent Unscented
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.