missing translation for 'onlineSavingsMsg'
Learn More

Merck Opticap™ Sterile XL300 Capsule Filters

For sterilizing grade performance and mycoplasma removal

Brand:  Merck KHVES003HH3

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 17888155

  • 880.00 € / Pack of 3
Estimated Shipment: 16-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • High-flux hydrophilic sterilizing-grade 0.1μm rated PES membrane for improved process economy
  • On-board PES membrane prefilter for extended throughput in high fouling media
  • Robust construction
  • Withstand multiple sterilization cycles
  • Easy wetting and integrity testing
  • Broad chemical compatibility across a wide pH range (pH 1–14)
  • Hydrophilic Polyethersulfone filter
  • Polyester support

  • Structural Components: Gamma stable polypropylene/Polysulfone
  • Silicone vent O-rings
  • Gamma compatible sterilization

Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals

Specifications

Specifications

0.5/0.1 μm
0.03 m2
0.56 in. (14 mm) Hose Barb
4.7 in.
Sterile
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
0.56 in. (14mm) Hose Barb
11.9 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers