missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Optifit Pipette Tips

Product Code. 31940822 Shop All Sartorius Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 960
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
31940822 Non-sterile Pack of 960
31940825 Sterile Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31940822 Supplier Sartorius Supplier No. LHG790200
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Minimizes the need for conventional materials while maintaining pipetting performance

  • Manufactured from virgin polypropylene.
  • Strict quality standards (ISO 9001 and ISO 13485) are followed at every step of the process, from RD to production and delivery.
  • The tips are manufactured in ISO 8 classified cleanroom conditions to avoid any contamination and are certified and tested for DNase, RNase, human DNA and endotoxins.
  • Presterilized tips are also available for laboratory work where purity requirements are of critical importance and are released for sale once they have fulfilled the test requirements.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Transparent
Autoclavable Not Autoclavable
Format Rack, 10 x 96
Tip Style Ergonomic
For Use With (Equipment) Sartorius
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.