missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Optima™ 5 High Performance GC Capillary Column

Brand:  Macherey-Nagel™ 726525.25

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10631302

  • 730.00 € / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0.53 mm
High Performance GC Capillary Column
Optima™ 5
340°C
25 m
G27
0.8 mm
Gas Chromatography (GC)
2 μm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers