missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Optima™ 5 Performance GC Capillary Column

Brand:  Macherey-Nagel™ 726325.50

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10012871

  • 1105.00 € / Pack of 50

Estimated Shipment: 28-06-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0.32 mm
High Performance GC Capillary Column
GC
340°C
50 m
1 Each
G27
0.5 mm
Optima™ 5
1 µm
Optima™ 5
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers