missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tray

Product Code. 10121863 Shop All Thermo Scientific Products
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10121863 Stainless Steel Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10121863 Supplier Thermo Scientific™ Supplier No. 50058672
  • ONLINE EXCLUSIVE PRICE
    353.00 €
    333.00 € / Pack of 2
    Save 20.00 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Material: Stainless Steel; For Use With: Heracell 150i

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
For Use With (Equipment) Heracell 150i
Material Stainless Steel
Type Tray
Width (English) 0.5 in.
Width (Metric) 1.27 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.