missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Set of Three Heratrays
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Heracell 240i
Unit Size:
Pack of 3
Description
Material: Stainless Steel; For Use With: Heracell 240i
Specifications
Specifications
| For Use With (Equipment) | Heracell 240i |
| Lighting | LED |
| Material | Stainless Steel |
| Type | Tray |
| Width (English) | 0.33 in. |
| Width (Metric) | 0.84 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction