Learn More
OPTIZYME™ Alkaline Phosphatase, Fisher BioReagents™
Description
OPTIZYME™ AP is a novel alkaline phosphatase that catalyzes the release of 5'- and 3'-phosphate groups from DNA, RNA and nucleotides in 10min.
Applications and Features:
- OPTIZYME™ Alkaline Phosphatase dephosphorylates cloning vector DNA (10min at 37°C)
- Fast thermoinactivation (5min at 75°C)
- Simultaneous digestion and dephosphorylation of vector DNA
- Single protocol for all types of DNA ends
- Nucleotide degradation prior to sequencing PCR products
- Active on nucleotides
- Protein dephosphorylation
- Active in all OPTIZYME restriction enzyme buffers
Supplied with:
10X OPTIZYME AP Buffer: 100mM Tris-HCl (pH 8.0 at 37°C), 50mM MgCl2, 1M KCl, 0.2% (v/v) Triton X-100, 1mg/mL BSA
Source: E. Coli cells with a cloned bacterial alkaline phosphatase gene
Quality Control:
- Endo-, exodeoxyribonuclease and ribonuclease-free
- Functionally tested for dephosphorylation of 5'-termini of overhanging, recessed and blunt DNA
Specifications
Specifications
| Concentration | 1 U/μL |
| Color | Colorless |
| pH | 7.4 |
| For Use With (Application) | DePhosphorylates cloning vector DNA (10 min. at 37°C), fast thermo-inactivation (5 min. at 75°C), Nucleotide degradation prior to sequencing PCR products, active on Nucleotides, Protein dephosphorylation, restriction En |
| Storage Buffer | OPTIZYME™ AP Buffer 10X : Tris HCl 100mM (8 pH at 37°C), MgCl2 50mM, KCl 1M, Triton™ X-100 0.2%, BSA 1mg/mL |
| Content And Storage | -20°C |
| Source | E. coli cells with a cloned bacterial Alkaline Phosphatase gene |
| Quantity | 3000 U |
| Product Type | Alkaline Phosphatase |
| Form | Solid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.