missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ ClaI (Bsu15I), Fisher BioReagents™

Product Code. 11914792
Click to view available options
Quantity:
2500 U
600 U
Concentration:
10 U/μL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11914792

Brand: Fisher BioReagents BP80241

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5'...A T^C G A T...3'
3'...T A G C^T A...5'

Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 20 - 50%
  • Buffer 2: 20 - 50%
  • Buffer 3: 20 - 50%
  • Buffer 4: 100%
  • Buffer 5: 20 - 50%
Storage Buffer:
  • 10mM Tris-HCl (pH 8.0 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/ml BSA and 50% (v/v) glycero
l
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after 50-fold over-digestion with ClaI.
  • Digestion of Agarose-embedded DNA:
  • A minimum of 5 units of ClaI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
  • Note:
    • ClaI is blocked by overlapping dam methylation. To avoid dam methylation, use a dam-, dcm- strain.
  • Compatible Ends: Bsp119I, Hin1I, Hin6I, HpaII, MaeII, MspI, NarI, Psp1406I, SsiI, TaqI, XmiI
  • Methylation Effects:
  • Dam: May overlap - blocked
    • 5'...ATCGm6A TC...3'
  • 3'...TAGC Tm6AG...5'
  • (Cleavage is blocked)
  • Dcm: Never overlaps - no effect
  • CpG: Completely overlaps - blocked
    • 5'...ATCGAT...3'
  • (Cleavage is blocked)
  • EcoKI: Never overlaps - no effect
  • EcoBI: May overlap - effect not determined
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Components >50% Water, 25 to 50% Glycerin
    Incubator Temperature 37°C
    pH 7.4
    For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Cfr9I
    Storage Buffer 10mM Tris HCl (pH 8 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 50% Glycerol
    Content And Storage Keep container tightly closed
    Quantity 600 U
    Cut Site AT.CGAT
    Product Type ClaI (Bsu15I)
    Form Liquid
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.