missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ DNA Polymerase I, Fisher BioReagents™

Product Code. 12817172
Change view
Click to view available options
Quantity:
2500 U
Unit Size:
2500 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12817172 2500 U 2500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12817172 Supplier Fisher BioReagents Supplier No. BP81095
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

OPTIZYME™ DNA Pol I is a template-dependent DNA Polymerase that catalyzes 5'->3' DNA synthesis, and exhibits 3'->5'-exonuclease, 5'->3' exonuclease and RNase H activity.

Applications and Features:

  • OPTIZYME DNA Polymerase I labels DNA by nick translation in conjunction with DNase I.
  • Second-strand cDNA synthesis in conjunction with RNase H.
  • Incorporation of modified nucleotides (e.g., biotin-, digoxigenin-, aminoallyl-, fluorescently-labeled nucleotides).
  • Active in buffers for OPTIZYME restriction enzymes and reverse transcriptases.

Supplied with:
10X OPTIZYME DNA Pol I Buffer: 500mM Tris-HCl (pH 7.5 at 25°C), 100mM MgCl2, 10mM DTT

Source: E. Coli cells with a cloned polA gene for E.Coli

Quality Control:
Endodeoxyribonuclease-free

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Color Colorless
pH 7.5
For Use With (Application) DNA Polymerase I labels DNA by nick translation in conjunction with DNase I, second-strand cDNA synthesis in conjunction with RNase H, incorporation of modified Nucleotides, Reverse Transcriptases
Storage Buffer OPTIZYME™ DNA Pol I Buffer 10X : Tris HCl 500mM (7.5 pH at 25°C), MgCl2 100mM, DTT 10mM
Content And Storage -20°C
Source E. coli cells with a cloned polA gene
Quantity 2500 U
Product Type DNA Polymerase I
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.