missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ DNase I (RNase-Free), Fisher BioReagents™

Product Code. 12887162
Change view
Click to view available options
Quantity:
1000 U
Unit Size:
1000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12887162 1000 U 1000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12887162 Supplier Fisher BioReagents Supplier No. BP81071
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

OPTIZYME™ DNase I is a recombinant endonuclease that digests DNA and is RNase-Free.

Applications and Features:

  • OPTIZYME DNase I, RNase-Free is used for the preparation of DNA-free RNA
  • Removal of template DNA following in vitro transcription
  • DNA labeling by nick translation in conjunction with DNA Polymerase I
  • Preparation of DNA-free RNA prior to RT-PCR
  • Studies of DNA-protein interactions by DNase I, RNase-free footprinting
  • Enzyme activity is strictly dependent on Ca2+ and is activated by Mg2+or Mn2+ions
  • Purified from non-animal host with a lower level of intrinsic RNases

Supplied with:

  • 10X OPTIZYME DNase I Buffer: 100mM Tris-HCl (pH 7.5 at 25°C), 25mM MgCl2, 1mM CaCl2
  • 50mM OPTIZYME EDTA (for inactivation by heat at 65°C for 10min)
Source: E. Coli cells with a cloned gene encoding bovine DNase I
Quality Control:
  • Ribonuclease-free
  • Functionally tested for the digestion of template DNA after in vitro transcription

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 U/μL
Color Colorless
pH 7.5
For Use With (Application) Preparation of DNA-free RNA, removal of template DNA following in-vitro transcription, DNA labeling by nick translation in conjunction with DNA Polymerase I, Preparation of DNA-free RNA prior to RT-PCR, Studies of DNA-Protein interactions by DNase I, RNase-free footprinting
Storage Buffer OPTIZYME™ DNase I Buffer 10X : Tris HCl 100mM (7.5 pH at 25°C), MgCl2 25mM, CaCl2 1mM, OPTIZYME™ EDTA 50mM (for inactivation by heat at 65°C for 10min.)
Content And Storage -20°C
Source E. coli cells with a cloned gene encoding bovine DNase I
Quantity 1000 U
Product Type DNase I (RNase-free)
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.