missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ DraI, Fisher BioReagents™
Description
5'...T T T^A A A...3'
3'...A A A^T T T...5'
Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
- Incubate at 37°C
- Buffer 1: 50 - 100%
- Buffer 2: 50 - 100%
- Buffer 3: 20 - 50%
- Buffer 4: 100%
- Buffer 5: 20 - 50%
- 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton X-100, 0.2mg/ml BSA and 50% (v/v) glycerol
- 5'...TTTAAm6AC(N)6G TGC...3'
Specifications
Specifications
| Concentration | 10 U/μL |
| Components | 10X OPTIZYME™ Buffer 4 |
| Incubator Temperature | 37°C |
| pH | 7.4 |
| For Use With (Application) | >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with DraI |
| Storage Buffer | 10mM Tris HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton™ X-100, 0.2mg/mL BSA, 50% Glycerol |
| Content And Storage | Keep container tightly closed |
| Quantity | 2000 U |
| Cut Site | TTT.AAA |
| Product Type | DraI |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?