missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ EcoRI, Fisher BioReagents™

Product Code. 11924842
Change view
Click to view available options
Quantity:
15,000 U
25,000 U
Concentration:
10 U/μL
50 U/μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Concentration unitSize
11924842 15,000 U 10 U/μL Each
11934812 25,000 U 50 U/μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11924842 Supplier Fisher BioReagents Supplier No. BP80035
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5'...G^A A T T C...3'
3'...C T T A A^G...5'

Conditions for 100% Activity:

  • 1X OPTIZYME Buffer EcoRI: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl, 0.02% Triton X-100 and 0.1mg/ml BSA.
  • Incubate at 37°C.
Supplied with: 10X OPTIZYME EcoRI Buffer
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 0 - 20%
  • Buffer 2: No reaction
  • Buffer 3: 100%
  • Buffer 4: No reaction
  • Buffer 5: 100%™
™star activity occurs at greater than 5-fold overdigestion (5 units X 1 hour)
Storage Buffer:
10mM potassium phosphate (pH 7.4 at 25°C), 300mM NaCl, 1mM EDTA, 1mM DTT, 0.2mg/ml BSA, 0.15% Triton X-100 and 50% (v/v) glycerol.
Ligation and Recleavage:
More than 95% of DNA fragments can be ligated and recut after a 50-fold overdigestion with EcoRI.
Digestion of Agarose-embedded DNA:
A minimum of 5 units of EcoRI is required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Note: Low salt concentration, large excess of the enzyme, pH>8.0, or the replacement of mg2+ by Mn2+ may result in star activity.
Compatible Ends: XapI, MunI, TasI.
Methylation Effects:
Dam: never overlaps - no effect.
Dcm: never overlaps - no effect.
CpG: may overlap - cleavage impaired.
  • 5'...m5C GAATTm5C G...3'
  • 3'... Gm5CTTAA Gm5...5'
  • EcoKI: never overlaps - no effect.
  • EcoBI: may overlap - no effect.
    • 5'...TGm6AATTC(N)4TGCT...3'
  • 3'...AC TTAAG(N)4m6ACGA...5'
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Components 25 to 50% Water, >50% Glycerin, 2.5 to10% Sodium Chloride
    Incubator Temperature 37°C
    pH 7.4
    For Use With (Application) >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with EcoRI
    Storage Buffer 10mM Potassium Phosphate (pH 7.4 at 25°C), 300mM NaCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 0.15% Triton™ X-100, 50% Glycerol
    Content And Storage Keep container tightly closed
    Quantity 15,000 U
    Cut Site G.AATTC
    Product Type EcoRI
    Form Liquid
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.