missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ M-MLV Reverse Transcriptase, Fisher BioReagents™

Product Code. 12837162
Change view
Click to view available options
Quantity:
25,000 U
5000 U
Unit Size:
25ku
5000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12837162 25,000 U 25ku
12827162 5000 U 5000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12837162 Supplier Fisher BioReagents Supplier No. BP81045
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

OPTIZYME™ M-MuLV RT is a genetically modified MLV RT that possesses RNA- and DNA-dependent polymerase activity.

Applications and Features:

  • OPTIZYME M-MLV RT is used for efficient first strand cDNA synthesis for RT-PCR and real-time RT-PCR (up to 13kb)
  • Generation of labeled cDNA probes for microarrays
  • Synthesis of cDNA for cloning and expression
  • Analysis of RNA by primer extension
  • DNA labeling
  • Optimum activity at 42°C (active up to 50°C)
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labeled nucleotides)

Supplied with:
5X OPTIZYME M-MLV RT Buffer: 250mM Tris-HCl (pH 8.3 at 25°C), 250mM KCl, 20mM MgCl2, 50mM DTT
Source: E. Coli cells with a cloned fragment of the pol gene encoding Moloney Murine Leukemia Virus reverse transcriptase
Quality Control:

  • Endo-, exodeoxyribonuclease, phosphatase and ribonuclease-free
  • Functionally tested in first strand cDNA and RT-PCR

TRUSTED_SUSTAINABILITY

Specifications

Concentration 200 U/μL
Color Colorless
pH 8.3
For Use With (Application) Generation of labeled cDNA probes for microarrays, synthesis of cDNA for cloning and expression, analysis of RNA by primer extension, DNA labeling, optimum activity at 42°C, incorporates modified Nucleotides
Storage Buffer OPTIZYME™ M-MLV RT Buffer 5X : Tris HCl 250mM (8.3 pH at 25°C), KCl 250mM, MgCl2 20mM, DTT 50mM
Content And Storage -20°C
Source E. coli cells with a cloned fragment of the pol gene encoding Moloney Murine Leukemia Virus Reverse Transcriptase
Quantity 25,000 U
Product Type OPTIZYME™ M-MLV Reverse Transcriptase
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.