missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ MspI (HpaII), Fisher BioReagents™

Product Code. 11974802
Change view
Click to view available options
Quantity:
3000 U
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11974802 3000 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11974802 Supplier Fisher BioReagents Supplier No. BP80481
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5'...C^C G G...3'
3'...G G C^C...5'

Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 50 - 100%
  • Buffer 2: 50 - 100%
  • Buffer 3: 0 - 20%
  • Buffer 4: 100%
  • Buffer 5: 0 - 20%
Storage Buffer:
  • 10mM potassium phosphate (pH 7.5 at 25°C), 200mM NaCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after 50-fold over-digestion with MspI.
  • Note:
    • When the external C in the sequence: CCGG is methylated, both MspI and HpaII cannot cleave. MspI is an isoschizomer of HpaII. However, unlike HpaII, MspI can cleave the sequence when the internal C residue is methylated.
  • Compatible Ends: Bsp119I, Bsu15I, Hin1I, Hin6I, MaeII, NarI, Psp1406I, SsiI, TaqI, XmiI
  • Methylation Effects:
  • Dam: Never overlaps - no effect
  • Dcm: Never overlaps - no effect
  • CpG: Completely overlaps - no effect
    • CCGG
  • (No effect on cleavage)
  • EcoKI: Never overlaps - no effect
  • EcoBI: Never overlaps - no effect
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Components 10X OPTIZYME™ Buffer 4
    Incubator Temperature 37°C
    pH 7.4
    For Use With (Application) Gene cloning and DNA genotyping
    Storage Buffer 10mM Potassium Phosphate (pH 7.5 at 25°C), 200mM NaCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
    Quantity 3000 U
    Cut Site C.CGG
    Product Type MspI
    Form Liquid
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.