missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ NdeI, Fisher BioReagents™

Product Code. 11964782
Click to view available options
Quantity:
1500 U
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11964782

Brand: Fisher BioReagents BP80201

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5'...C A^T A T G...3'
3'...G T A T^A C...5'

Supplied with: 10X OPTIZYME Buffer 3
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 3: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 0 - 20%
  • Buffer 2: 0 - 20%
  • Buffer 3: 100%
  • Buffer 4: 0 - 20%
  • Buffer 5: 50 - 100%
Storage Buffer:
  • 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after 50-fold over-digestion with NdeI.
  • Digestion of Agarose-embedded DNA:
  • A minimum of 5 units of NdeI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
  • Compatible Ends: Csp6I, FspBI, Tru1I, VspI
  • Methylation Effects:
  • Dam: Never overlaps - no effect
  • Dcm: Never overlaps - no effect
  • CpG: Never overlaps - no effect
  • EcoKI: Never overlaps - no effect
  • EcoBI: May overlap - no effect
    • 5'...TGm6A(N)4CATA TGCT...3'
  • 3'...AC T(N)4GTATm6ACGA...5'
  • (No effect on cleavage)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Components 25 to 50% Water, >50% Glycerin
    pH 7.4
    For Use With (Application) >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Nde I
    Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
    Content And Storage Keep container tightly closed
    Quantity 1500 U
    Cut Site CA.TATG
    Product Type NdeI
    Form Liquid
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.