missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ PfoI, Fisher BioReagents™

Product Code. 11924832
Change view
Click to view available options
Quantity:
200 U
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11924832 200 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11924832 Supplier Fisher BioReagents Supplier No. BP80771
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5'...T^C C N G G A...3'
3'...A G G N C C^T...5'

Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 0 - 20%
  • Buffer 2: 20 - 50%
  • Buffer 3: 50 - 100%
  • Buffer 4: 100%
  • Buffer 5: 0 - 20%
Storage Buffer:
  • 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after 50-fold over-digestion with PfoI.
  • Digestion of Agarose-embedded DNA:
  • A minimum of 10 units of PfoI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
  • Note:
    • PfoI cleavage is impaired by overlapping dam methylation and blocked by overlapping dcm methylation. To avoid dam or dcm methylation, use a dam-, dcm- strain.
  • Compatible Ends: EcoRII, SexAI, BssKI
  • Methylation Effects:
  • Dam: May overlap - cleavage impaired
    • 5'...TCCNGGm6A TC...3'
  • 3'...AGGNCC Tm6AG...5'
  • (Cleavage is impaired)
  • Dcm: May overlap - blocked
    • 5'...TCm5CA GGA...3'
  • 3'...AG GTm5CCT...5'
  • (Cleavage is blocked)
  • CpG: May overlap - blocked
    • 5'...TCm5C GGGA...3'
  • 3'...AG Gm5CCCT...5'
  • (Cleavage is blocked)
  • EcoKI: Never overlaps - no effect
  • EcoBI: Never overlaps - no effect
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Components >50% Glycerin, 25 to 50% Water
    Incubator Temperature 37°C
    pH 7.4
    For Use With (Application) >90% of DNA fragments can be ligated and re-cut after 10-fold over-digestion with Pfo I
    Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
    Content And Storage Keep container tightly closed
    Quantity 200 U
    Cut Site T.CCNGGA
    Product Type PfoI
    Form Liquid
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.