missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ SacI, Fisher BioReagents™
Description
5'...G A G C T^C...3'
3'...C^T C G A G...5'
Supplied with: 10X OPTIZYME Ecl136II, SacI Buffer
Conditions for 100% Activity:
- 1X OPTIZYME Buffer Ecl136II, SacI: 10mM Bis-Tris Propane-HCl (pH 6.5 at 37°C), 10mM MgCl2 and 0.1mg/ml BSA
- Incubate at 37°C
- Buffer 1: 50 - 100%
- Buffer 2: 20 - 50%
- Buffer 3: 0 - 20%
- Buffer 4: 50 - 100%
- Buffer 5: 0 - 20%
- 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1 mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
- SacI is sensitive to cytosine methylation at GAGmCTC but not GAGCTmC and insensitive to adenine methylation at GmAGCTC/. AluI methyltransferase (AGmCT) can be used to block SacI.
- Supercoiled plasmids may require up to 5-fold more SacI for complete digestion than linear DNA.
- SacI is inhibited by common clinical anticoagulants found in some preparation of anticoagulated peripheral blood and bone marrow. Levels of EDTA and ACD (citric acid-sodium citrate-dextrose) in standard sample preparation have been shown to inhibit SacI. Three times the normal concentration for heparin is required to inhibit SacI.
Methylation Effects:
Dam: Never overlaps - no effect
Dcm: Never overlaps - no effect
CpG: May overlap - no effect
- 5'...m5C GAGCTm5C G...3'
- 5'...TGm6AGCTC(N)4 TGCT...3'
Specifications
Specifications
| Concentration | 10 U/μL |
| Components | 10X OPTIZYME™ Ecl136II, SacI Buffer |
| Incubator Temperature | 37°C |
| pH | 7.4 |
| For Use With (Application) | >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with SacI |
| Storage Buffer | 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol |
| Quantity | 2000 U |
| Cut Site | GAGCT.C |
| Product Type | SacI |
| Form | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?