missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ Sau3AI (Bsp143I), Fisher BioReagents™

Product Code. 11954792
Change view
Click to view available options
Quantity:
300 U
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11954792 300 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11954792 Supplier Fisher BioReagents Supplier No. BP80301
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5'...^G A T C ...3'
3'... C T A G^...5'

Supplied with: 10X OPTIZYME Buffer Sau3AI
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer Sau3AI: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate, 0.02% Triton X-100 and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 20 - 50%
  • Buffer 2: 20 - 50%
  • Buffer 3: 0 - 20%
  • Buffer 4: 50 - 100%
  • Buffer 5: 0 - 20%
Storage Buffer:
  • 10mM Tris-HCl (pH 7.4 at 25°C), 10mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after 50-fold over-digestion with Sau3AI
  • Compatible Ends: BamHI, BclI, BglII, PsuI
  • Methylation Effects:
  • Dam: Completely overlaps - no effect
    • 5'...Gm6ATC...3'
  • (No effect on cleavage)
  • Dcm: Never overlaps - no effect
  • CpG: May overlap - blocked
    • 5'...GATm5C G...3'
  • 3'...CTA Gm5C...5'
  • (Cleavage is blocked)
  • EcoKI: Never overlaps - no effect
    • 5'...TGm6ATC(N)6 TGCT...3'
  • 3'...AC TAG(N)6 m6ACGA...5'
  • (No effect on cleavage)
  • EcoBI: May overlap - no effect
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Components 10X OPTIZYME™ Buffer Sau3AI
    Incubator Temperature 37°C
    pH 7.4
    For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Sau3AI
    Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol
    Quantity 300 U
    Cut Site .GATC.
    Product Type Sau3AI
    Form Liquid
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.