missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ T3 RNA Polymerase, Fisher BioReagents™
Description
OPTIZYME™ T3 RNA Pol is a DNA-dependent RNA polymerase that catalyzes the 5'->3' synthesis of RNA from DNA.
Applications and Features:
- OPTIZYME™ T3 RNA Polymerase synthesizes unlabeled and labeled RNA that can be used for: (1) hybridization, (2) in vitro RNA translation, (3) RNase protection assays, (4) genomic DNA sequencing, (5) RNA splicing, and (6) studies of RNA secondary structure and RNA-protein interactions
- Incorporation of modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Supplied with:
5X OPTIZYME Transcription Buffer: 200mM Tris-HCl (pH 7.9 at 25°C), 30mM MgCl2, 50mM DTT, 50mM NaCl, 10mM spermidine
Source: E. Coli cells with a cloned T3 RNA Polymerase gene
Quality control:
- Endo-, exodeoxyribonuclease and ribonuclease-free
- Functionally tested in in vitro transcription reaction
Specifications
Specifications
| Concentration | 20 U/μL |
| Color | Colorless |
| pH | 7.5 |
| For Use With (Application) | Hybridization, in-vitro RNA translation, RNase protection assay, genomic DNA sequencing, RNA splicing and studies of RNA secondary structure and RNA-Protein interaction, quality control |
| Storage Buffer | OPTIZYME™ Transcription Buffer 5X : Tris HCl 200mM (7.9 pH at 25°C), MgCl2 30mM, DTT 50mM, NaCl 50mM, Spermidine 10mM |
| Content And Storage | -20°C |
| Source | E. coli cells with a cloned T3 RNA Polymerase gene |
| Quantity | 1000 U |
| Product Type | T3 RNA Polymerase |
| Form | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?