missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ XbaI, Fisher BioReagents™
Description
5'...T^C T A G A...3'
3'...A G A T C^T...5'
Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity
- 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
- Incubate at 37°C
- Buffer 1: 50 - 100%
- Buffer 2: 50 - 100%
- Buffer 3: 20 - 50%
- Buffer 4: 100%
- Buffer 5: 0 - 20%
10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 7mM 2-mercaptoethanol, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
Ligation and Re-cleavage:
More than 95% of the DNA fragments can be ligated and re-cut after a 50-fold over-digestion with XbaI.
Digestion of Agarose-embedded DNA:
A minimum 5 units of XbaI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Notes:
- XbaI is blocked by overlapping dam methylation.
- To avoid dam methylation, use a dam-, dcm- strain.
Methylation Effects:
Dam: May overlap - blocked
- 5'...TCTAGm6A TC...3'
- 3'...AGATC Tm6AG...5'
- (Cleavage is blocked)
CpG: Never overlaps - no effect
EcoKI: Never overlaps - no effect
EcoBI: Never overlaps - no effect
Specifications
Specifications
| Concentration | 10 U/μL |
| Components | 10X OPTIZYME™ Buffer 4 |
| Incubator Temperature | 37°C |
| pH | 7.4 |
| For Use With (Application) | Indicates ligation efficiency of DNA fragments generated by digestion with the Restriction Enzyme |
| Storage Buffer | 10mM Tris HCl (pH 7.4), 100mM KCl, 7mM 2-mercaptoethano, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol |
| Quantity | 2000 U |
| Cut Site | T.CTAGA |
| Product Type | XbaI |
| Form | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?