missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ XbaI, Fisher BioReagents™

Product Code. p-4362858
Click to view available options
Quantity:
10,000 U
2000 U
Concentration:
10 U/μL
50 U/μL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11994842

Brand: Fisher BioReagents BP80085

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5'...T^C T A G A...3'
3'...A G A T C^T...5'

Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity

  • 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 50 - 100%
  • Buffer 2: 50 - 100%
  • Buffer 3: 20 - 50%
  • Buffer 4: 100%
  • Buffer 5: 0 - 20%
Storage Buffer:
10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 7mM 2-mercaptoethanol, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
Ligation and Re-cleavage:
More than 95% of the DNA fragments can be ligated and re-cut after a 50-fold over-digestion with XbaI.
Digestion of Agarose-embedded DNA:
A minimum 5 units of XbaI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Notes:
  • XbaI is blocked by overlapping dam methylation.
  • To avoid dam methylation, use a dam-, dcm- strain.
Compatible Ends: BcuI, Eco130I, NheI, XmaJI
Methylation Effects:
Dam: May overlap - blocked
  • 5'...TCTAGm6A TC...3'
  • 3'...AGATC Tm6AG...5'
  • (Cleavage is blocked)
Dcm: Never overlaps - no effect
CpG: Never overlaps - no effect
EcoKI: Never overlaps - no effect
EcoBI: Never overlaps - no effect

TRUSTED_SUSTAINABILITY

Specifications

Concentration 50 U/μL
Components 10X OPTIZYME™ Buffer 4
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Indicates ligation efficiency of DNA fragments generated by digestion with the Restriction Enzyme
Storage Buffer 10mM Tris HCl (pH 7.4), 100mM KCl, 7mM 2-mercaptoethano, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Quantity 10,000 U
Cut Site T.CTAGA
Product Type XbaI
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.