missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Optochin Disc

Product Code. 11429858
Change view
Click to view available options
Packaging Type:
1 Cartridge/Pk.
5 Cartridges/Pk.
Unit Size:
Pack of 250
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Type Unit Size
11429858 5 Cartridges/Pk. Pack of 250
11903842 1 Cartridge/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429858 Supplier Thermo Scientific™ Supplier No. DD0001B
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Oxoid Optochin Disc is for the differentiation of Streptococcus pneumoniae.

Thermo Scientific™ Oxoid Optochin Disc is for the differentiation of Streptococcus pneumoniae.

Ordering Alerts:

5μg. 50 Discs/Cartridge. 1 Cart/Pk.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Optochin (Ethylhydrocuprein Hydrochloride)
Concentration 5 μg
Product Type Optochin Disc
Test Type Microbial Differentiation
Material Paper
Packaging Type 5 Cartridges/Pk.
Quantity 50 Discs/Cartridge
Species Streptococcus pneumoniae
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.